Pinometostat (EPZ-5676) 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(((cis-3-(2-(5-(tert-butyl)-1H-benzo[d]imidazol-2-yl)ethyl)cyclobutyl)(isopropyl)amino)methyl)tetrahydrofuran-3,4-diol, Pinometostat

468161000
ACRO468161000EA 553 EUR
ACRO468161000
Pinometostat (EPZ-5676) 95%
Pinometostat (EPZ-5676)

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₃₀H₄₂N₈O₃
Molecular Weight: 562.71 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD24849416
CAS-Nummer: 1380288-87-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR