EPZ-6438 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Tazemetostat

468171000
ACRO468171000EA 157 EUR
ACRO468171000
EPZ-6438 95%
Tazemetostat

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₃₄H₄₄N₄O₄
Molecular Weight: 572,74 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD20527867
CAS-Nummer: 1403254-99-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR