Filgotinib (GLPG0634) 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(5-(4-((1,1-Dioxidothiomorpholino)methyl)phenyl)-[1,2,4]triazolo[1,5-a]pyridin-2-yl)cyclopropanecarboxamide, Filgotinib

468181000
ACRO468181000EA 179 EUR
ACRO468181000
Filgotinib (GLPG0634) 95%
Filgotinib (GLPG0634)

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₁H₂₃N₅O₃S
Molecular Weight: 425,5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD20527867
CAS-Nummer: 1206161-97-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR