GNF-5 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N-(2-Hydroxyethyl)-3-(6-((4-(trifluoromethoxy)phenyl)amino)pyrimidin-4-yl)benzamide, N-(2-hydroxyethyl)-3-[6-[4-(trifluoromethoxy)anilino]pyrimidin-4-yl]benzamide

468191000
ACRO468191000EA 405 EUR
ACRO468191000
GNF-5 95%
GNF-5

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₀H₁₇F₃N₄O₃
Lagertemperatur: Gefrierschrank
MDL: MFCD16877246
CAS-Nummer: 778277-15-9
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR