GSK481 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-5-Benzyl-N-(5-methyl-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]oxazepin-3-yl)isoxazole-3-carboxamide, 5-(Phenylmethyl)-N-[(3S)-2,3,4,5-tetrahydro-5-methyl-4-oxo-1,5-benzoxazepin-3-yl]-3-isoxazolecarboxamide

468201000
ACRO468201000EA 448 EUR
ACRO468201000
GSK481 95%
GSK481

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₁H₁₉N₃O₄
Molecular Weight: 377,4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD30343843
CAS-Nummer: 1622849-58-4

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or crystalline powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR