UNC1999 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Isopropyl-6-(6-(4-isopropylpiperazin-1-yl)pyridin-3-yl)-N-((6-methyl-2-oxo-4-propyl-1,2-dihydropyridin-3-yl)methyl)-1H-indazole-4-carboxamide

468221000
ACRO468221000EA 563 EUR
ACRO468221000
UNC1999 95%
UNC1999

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₃₃H₄₃N₇O₂
Molecular Weight: 569,7402 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD26960958
CAS-Nummer: 1431612-23-5

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or crystalline powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR