ABT-199 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: GDC-0199, Venetoclax, 2-((1H-Pyrrolo[2,3-b]pyridin-5-yl)oxy)-4-(4-((4'-chloro-5,5-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((3-nitro-4-(((tetrahydro-2H-pyran-4-yl)methyl)amino)phenyl)sulphonyl)benzamide

468231000
ACRO468231000EA 131 EUR
ACRO468231000
ABT-199 95%
ABT-199

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₄₅H₅₀ClN₇O₇S
Molecular Weight: 868,44 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD23160052
CAS-Nummer: 1257044-40-8

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR