P505-15 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: PRT062607

468261000
ACRO468261000EA 279 EUR
ACRO468261000
P505-15 95%
P505-15

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₁₉H₂₃N₉O
Molecular Weight: 393,45 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11878503
CAS-Nummer: 1370261-96-3

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR