SCH772984 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
SCH-772984, (R)-1-(2-Oxo-2-(4-(4-(pyrimidin-2-yl)phenyl)piperazin-1-yl)ethyl)-N-(3-(pyridin-4-yl)-1H-indazol-5-yl)pyrrolidine-3-carboxamide, (3R)-1-[2-oxo-2-[4-(4-pyrimidin-2-ylphenyl)piperazin-1-yl]ethyl]-N-(3-pyridin-4-yl-1H-indazol-5-yl)pyrrolidine-3-carboxamide, SCH 772984, SCH772984
Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.
Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies
Formel:
C₃₃H₃₃N₉O₂ Molecular Weight: 587,67 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD28144700 CAS-Nummer: 942183-80-4 |
Spezifikation Testergebnisse
Appearance | Yellow Powder or solid |
HPLC | ≥94% |
MS | complies |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...