Favipiravir

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: T-705

468691000
ACRO468691000EA 150 EUR
ACRO468691000
Favipiravir
Favipiravir
Formel: C₅H₄FN₃O₂
Molecular Weight: 157.09 g/mol
Siedepunkt: 52.6°C (760mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD16879084
CAS-Nummer: 259793-96-9

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥ 96.0%
Water ≤ 2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR