Multi-element standard solutions for ion chromatography
Lieferant: CPACHEM
1521.1K.W.L1
1521.1K.W.L25
1521.K1.W.L1
1521.K1.W.L25
ACSD1521.1K.W.L1EA
177
EUR
ACSD1521.1K.W.L1
ACSD1521.1K.W.L25
ACSD1521.K1.W.L1
ACSD1521.K1.W.L25
Multi-element standard solutions for ion chromatography
Standards
Anionische Standards
Anionische Multi-Element-Standards
This standard solution is intended for ion chromatography.
Multi element standards for Br⁻, Cl-, F⁻, NO₂⁻, NO₃⁻, PO₄³⁻, SO₄²⁻ diluted in H₂O.