Ammoniaklösung, Testlösung (TS)

Lieferant: CPACHEM

Synonyme: Ammoniumhydroxid, Ammoniak

CH8002013.1L
ACSDCH8002013.1LEA 75 EUR
ACSDCH8002013.1L
Ammoniaklösung, Testlösung (TS)
Ammoniaklösung

Standards: ISO 9001 certified.

Formel: NH₃
Lagertemperatur: Room Temperature
MDL: MFCD00066650
CAS-Nummer: 1336-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR