Eriochromschwarz T-Gemisch Indikator

Lieferant: CPACHEM

CH8005015.L04
ACSDCH8005015.L04EA 50.5 EUR
ACSDCH8005015.L04
Eriochromschwarz T-Gemisch Indikator
Eriochromschwarz T

Standards: ISO 9001

Formel: C₂₀H₁₂N₃NaO₇S
Schmelzpunkt: 801 °C
Lagertemperatur: Room Temperature
MDL: MFCD00003935
CAS-Nummer: 1787-61-7

Bestellen

Spezifikation Testergebnisse

Content 0.1g of methyl red in 7.4ml of 0.05mol/l NaOH solution and diluted with water to 200ml.
Performance Test passed
Colour change from red to yellow (pH 4.2 - 6.3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR