Methylrot-Lösung Indikator

Lieferant: ACTIVE MOTIF

Synonyme: 2-(4-(Dimethylamino)phenylazo)benzoesäure

CH8005022.L04
ACSDCH8005022.L04EA 50.5 EUR
ACSDCH8005022.L04
Methylrot-Lösung Indikator
Methylrot

Standards: ISO 9001

Formel: C₁₅H₁₅N₃O₂
Molecular Weight: 269,3 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00002425
CAS-Nummer: 493-52-7

Bestellen

Spezifikation Testergebnisse

Content 0.1g of methyl red in 7.4ml of 0.05mol/l NaOH solution and diluted with water to 200ml.
Performance Test passed
Colour change from red to yellow (pH 4.2 - 6.3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR