Bismut(III)nitrat 0,01 mol/L

Lieferant: ACTIVE MOTIF

Synonyme: Bismuth trinitrate

CH8006005.1L
ACSDCH8006005.1LEA 95.1 EUR
ACSDCH8006005.1L
Bismut(III)nitrat 0,01 mol/L
Bismut(III)nitrat

Standards: ISO 9001 certified.


Bestellinformation: Shelf life on ship date: 12 months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.

Formel: Bi(NO₃)₃·xH₂O
Molecular Weight: 394,99 g/mol (anhy)
Siedepunkt: 100 °C
Dichte: 1 g/cm³
MDL: MFCD00003460
CAS-Nummer: 10361-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR