System Calibration Standard (7 components)
Lieferant: CPACHEM
F237594
ACSDF237594EA
493
EUR
ACSDF237594
System Calibration Standard (7 components)
Standards
Mineralölindustrie-Standards
System calibration standard with 7 components.
- Solvent: n-Heptane
- Volume: ampoule 5 ml
The components consists of cyclohexane 1% (w/v), 1-phenyldodecane 0,1% (w/v), o-Xylene (1,2-dimethylbenzene) 0.5% (w/v), hexamethylbenzene 0,1% (w/v), naphthalene 0,1% (w/v), dibenzothiophene 0,05% (w/v) 9-methylanthracene 0,05% (w/v).
Standards: EN 12916