Magnesium Standard Solution, 10000 mg/L Mg in dil. Nitric Acid Standard for ICP-MS
Lieferant: ACTIVE MOTIF
M132.2NP.L1
ACSDM132.2NP.L1EA 231 EUR
ACSDM132.2NP.L1
Magnesium Standard Solution, 10000 mg/L Mg in dil. Nitric Acid Standard for ICP-MS
Standards Kationischer Standards Magnesium-Standards
Mg 10000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Standards: ISO 17034, ISO 17025, ISO 9001.