Silver Standard Solution, 1000 mg/L Ag in dil. Nitric Acid Standard for ICP-MS
Lieferant: CPACHEM
M201.2NP.L1
ACSDM201.2NP.L1EA
123
EUR
ACSDM201.2NP.L1
Silver Standard Solution, 1000 mg/L Ag in dil. Nitric Acid Standard for ICP-MS
Standards
Kationischer Standards
Silber-Standards
Ag 1000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Standards: ISO 17034, ISO 17025, ISO 9001.