Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid Standard for ICP-MS
Lieferant: ACTIVE MOTIF
M208.2NP.L1
ACSDM208.2NP.L1EA
123
EUR
ACSDM208.2NP.L1
Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid Standard for ICP-MS
Standards
Kationischer Standards
Wismut-Standards
Bi 1000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.