Terbium Standard Solution, 1000 mg/L Tb in dil. Nitric Acid Standard for ICP-MS
Lieferant: ACTIVE MOTIF
M258.2NP.L1
ACSDM258.2NP.L1EA
159
EUR
ACSDM258.2NP.L1
Terbium Standard Solution, 1000 mg/L Tb in dil. Nitric Acid Standard for ICP-MS
Standards
Kationischer Standards
Terbium-Standards
Tb 1000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Standards: ISO 17034, ISO 17025, ISO 9001.