Natriumbromid 99.99%, HighPurS-CERT
Lieferant: ACTIVE MOTIF
Synonyme:
Bromnatrium
PSNABRC.50G
ACSDPSNABRC.50GEA
513
EUR
ACSDPSNABRC.50G
Natriumbromid 99.99%, HighPurS-CERT
Natriumbromid
Bestellinformation: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Formel:
NaBr Molecular Weight: 102,89 g/mol Siedepunkt: 1,39 °C Schmelzpunkt: 747 °C Dichte: 0,0032 g/cm³ |
MDL:
MFCD00003475 CAS-Nummer: 7647-15-6 EINECS: 231-599-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...