Natriumnitrat 99.99%, HighPurS-CERT
Lieferant: CPACHEM
|
Gefahr
|
Synonyme:
Natronsalpeter
PSNANAC.50G
ACSDPSNANAC.50GEA
187
EUR
ACSDPSNANAC.50G
Natriumnitrat 99.99%, HighPurS-CERT
Natriumnitrat
Standards: ISO 17034, ISO 17025, ISO 9001.
Bestellinformation: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Formel:
NaNO₃ Siedepunkt: 600,..700 °C Schmelzpunkt: 312 °C Dichte: 2,26 g/cm³ |
MDL:
MFCD00011119 CAS-Nummer: 7631-99-4 EINECS: 231-554-3 UN: 1498 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...