Analytical Standard, Nicarbazin, Reference Material
Lieferant: CPACHEM
SB18251.100MG
ACSDSB18251.100MGEA 169 EUR
ACSDSB18251.100MG
Analytical Standard, Nicarbazin, Reference Material
Standards Organische Standards
Bestellinformation: Shelf life on ship date: 24 months - store in a refrigerator at temperatures between 2 to 8 °C.