Cumol
Lieferant: ACTIVE MOTIF
|
Gefahr
|
Synonyme:
2-Phenylpropan, Isopropylbenzol
SB224.500MG
ACSDSB224.500MGEA
70.6
EUR
ACSDSB224.500MG
Cumol
Cumol
Shelf Life on ship date: 24 months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Formel:
C₉H₁₂ Molecular Weight: 120,19 g/mol Siedepunkt: 152 °C Schmelzpunkt: –96,9 °C Dichte: 0,86 g/cm³ Flammpunkt: 36 °C |
MDL:
MFCD00008881 CAS-Nummer: 98-82-8 EINECS: 202-704-5 UN: 1918 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...