Dimetridazol
Lieferant: ACTIVE MOTIF
|
Achtung
|
Synonyme:
1,2-Dimethyl-5-nitroimidazole
SB32100.250MG
ACSDSB32100.250MGEA
71.2
EUR
ACSDSB32100.250MG
Dimetridazol
Dimetridazol
Formel:
C₅H₇N₃O₂ Molecular Weight: 141,13 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00047046 CAS-Nummer: 551-92-8 EINECS: 209-001-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...