Decoquinate

Lieferant: ACTIVE MOTIF

Synonyme: Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester

SB50590.100MG
ACSDSB50590.100MGEA 262 EUR
ACSDSB50590.100MG
Decoquinate
Decoquinate
Formel: C₂₄H₃₅NO₅
Molecular Weight: 417,55 g/mol
Schmelzpunkt: 242,..246 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00673686
CAS-Nummer: 18507-89-6
EINECS: 242-389-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR