Certified Reference Material for Standardisation of Volumetric Solutions
Lieferant: ACTIVE MOTIF
VS66332.80G
ACSDVS66332.80GEA 572 EUR
ACSDVS66332.80G
Certified Reference Material for Standardisation of Volumetric Solutions
Standards Organische Standards
Iron(II)ethylenediammonium sulfate tetrahydrate - certified reference material for standardisation of volumetric solutions.
Shelf Life on Ship Date: 60 months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Standards: ISO 17034, ISO 17025, ISO 9001.