Liquid viewer, high intensity, Apollo II
Lieferant: ADELPHI MANUFACTURING
6031520
ADLM6031520EA 0 EUR
ADLM6031520
Liquid viewer, high intensity, Apollo II
Beleuchtung Leuchtkästen
Designed for use in pharmaceutical and laboratory environments, the Apollo II Liquid Viewer for coloured or opaque glass containers provides a simple method for the detection of visible particles in ampoules, vials, and bottles.
  • Height and angle adjustable
  • Intuitive assembly; no tools required
  • Light level between 8,000 and 10,000 Lux
  • Unit can fold for convenient storage, with the inspection surfaces folding together to reduce chance of damage
The unit is packaged in sustainably sourced foam, made from polyethylene resin which is derived from renewable sugarcane-based feedstock.
The Apollo II High Intensity Liquid Viewer is suitable for coloured glass containers. Adelphi Manufacturing also offer the Apollo II Liquid Viewer for transparent glass containers, and the Apollo I Liquid Viewer, with additional magnifier.
Standards: This unit conforms to the European Pharmacopoeia Specification 2.9.20 and to the US Pharmacopoeia – USP chapter 41 (1790), section 6.1.
Adelphi Manufacturing’s ISO 9001:2015 certification guarantees the high quality finish of this product. Supporting IQ/OQ documentation can be supplied. User manuals are available in English, Spanish, French, German, Dutch and Czech – additional languages are also available upon request.