Syringe Stopper Press
Lieferant: ADELPHI MANUFACTURING
SP048-3001
ADLMSP048-3001EA 0 EUR
ADLMSP048-3001
Syringe Stopper Press
Pressen
Easy and efficient insertion of stoppers into pre-filled syringes of 6,35 mm bore or greater.
  • Easy and efficient insertion of stoppers into pre-filled syringes, ensuring that all air is expelled
  • Hygienic 316 L pharmaceutical grade stainless steel contact parts, to eliminate the risk of contamination
  • Fully adjustable design for operator comfort and safety
The unit comprises a syringe holding jig, lever operated stopper press, and a divide for aiding the removal of air from the syringe.
Bestellinformation: Adelphi also offers the Accuramatic peristaltic pump (purchased separately), for filling nested syringes.