Vacuum microanalysis holders
Lieferant: Advantech
311110
ADVT311110EA 428 EUR
ADVT311110
Vacuum microanalysis holders
Trichter Filtertrichter Mikro-Filtertrichter
Glass Microanalysis Holders 13 mm.
  • Concentrate samples in a small area (1,2 cm²)
  • KG 13 AA features a two-part funnel (13 ml/100 ml)
  • Filters small volume of liquid