Thermocouple input module
Lieferant: ADVANTEC
ADAM-5018-A1E
ADVTADAM-5018-A1EEA 0 EUR
ADVTADAM-5018-A1E
Thermocouple input module
Temperaturfühler
Thermocouple with seven channels input module that features programmable input ranges on all channels.
It accepts millivolt inputs (±15 mV, ±50 mV, ±100 mV, ±500 mV) and voltage inputs upto (±1V, ±2,5 V).
The module forwards the data to the host computer in engineering units (mV, V, mA or temperature °C). An external CJC on the plug-in terminal is designed for accurate temperature measurement.