Membrane filters, hydrophilic, PTFE
Lieferant: ADVANTEC
H020A025A
ADVTH020A025AEA 314 EUR
ADVTH020A025A
Membrane filters, hydrophilic, PTFE
Filter Membranfilter
Hydrophilic PTFE membrane filters are ideal for HPLC and other mixtures of aqueous and organic solvents.
  • Chemical and pH resistance
  • Optically clear when wet with water
  • High flow rates with minimal aqueous extractables
  • Not autoclavable