Dissecting scissors, Aesculap® Durotip®
Lieferant: Aesculap
BC258R BC259R BC259W
AESCBC258REA 190 EUR
AESCBC258R AESCBC259R AESCBC259W
Dissecting scissors, Aesculap® Durotip®
Scheren
Dissecting scissor for dental applications. Type Baby-Metzenbaum.
  • With inserted carbide cutting edges
  • Excellent cutting accuracy
  • Long working life
  • Gold-plated rings
Wave cut models prevent tissue from slipping out during the cutting process.