Tracheal retractors, Aesculap®
Lieferant: Aesculap
BT106R BT107R BT103R
AESCBT106REA 0 EUR
AESCBT106R AESCBT107R AESCBT103R
Tracheal retractors, Aesculap®
Retraktoren
These handheld retractors are used during tracheotomy procedures. They are available as a single or multi-prong instrument where the prong(s) hold back cricoid cartilage prior to the tracheal incision.