Gum scissors, Aesculap® Durotip®
Lieferant: Aesculap
DO208R DO209R DO250R
AESCDO208REA 0 EUR
AESCDO208R AESCDO209R AESCDO250R
Gum scissors, Aesculap® Durotip®
Scheren
Gum scissors with inserted carbide cutting edges.
  • Excellent cutting accuracy
  • Long working life
  • Gold-plated rings
  • Three years warranty