Micro scissors for endoscopic work, Aesculap® PaediScope®
Lieferant: Aesculap
FF373R
AESCFF373REA 0 EUR
AESCFF373R
Micro scissors for endoscopic work, Aesculap® PaediScope®
Scheren
Flexible micro scissors for advanced intraventricular Neuroendoscopy.
Ø = 1 mm