Micro scissors for endoscopic work, Aesculap® MINOP®
Lieferant: Aesculap
FF385R FF386R FF432R FF433R FF435R FF436R
AESCFF385REA 0 EUR
AESCFF385R AESCFF386R AESCFF432R AESCFF433R AESCFF435R AESCFF436R
Micro scissors for endoscopic work, Aesculap® MINOP®
Scheren
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
  • Very delicate
Ø = 2 mm
Bestellinformation: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.