Suture, Dafilon (Aesculap®)
Lieferant: Aesculap
G118110
AESCG118110EA 0 EUR
AESCG118110
Suture, Dafilon (Aesculap®)
Fäden
Dafilon is a non-absorbable nylon monofilament suture.
  • Minimum tissue dragging
  • Inert material with good tissue compatibility
  • Excellent pliability
Suitable for skin closure in ophthalmic surgery, reconstructive surgery, and microsurgery.