Drill handpiece, Aesculap®
Lieferant: Aesculap
GB169R
AESCGB169REA 0 EUR
AESCGB169R
Drill handpiece, Aesculap®
Probenehmer Probenehmer für Feststoffe
Drill handpiece for exchangeable chucks. Features power and stability during skull perforations.
  • Proven versatility at low speed
  • Longer and slimmer design for additional visibility
Bestellinformation: Motor, control unit, spray nozzle and cables need to be ordered separately.
Lieferumfang: Includes a ring spanner.