Dental trays, stainless steel
Lieferant: Agencinox
1210 1215
AGEN1210EA 20.3 EUR
AGEN1210 AGEN1215
Dental trays, stainless steel
Schalen/Schüsseln/Töpfe Instrumentenschalen
Stainless steel, 18/10.
  • Smooth surfaces for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance