Instrument trays, rectangular
Lieferant: Agencinox
1252 1242
AGEN1252EA 182 EUR
AGEN1252 AGEN1242
Instrument trays, rectangular
Schalen/Schüsseln/Töpfe Instrumentenschalen
Stainless steel, 18/10.
  • Smooth surfaces for fast, easy cleaning
  • Good chemical resistance
  • Excellent thermal resistance