Instrument trays, with lid
Lieferant: Agencinox
5029 5036 5046 5052 5075
AGEN5029EA 93 EUR
AGEN5029 AGEN5036 AGEN5046 AGEN5052 AGEN5075
Instrument trays, with lid
Schalen/Schüsseln/Töpfe Instrumentenschalen
Stainless steel, non-rusting.
  • Special corner shape for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance
  • Excellent thermal resistance
With a cover that enables the instrument dishes to be stacked.