Trolley, trays
Lieferant: Agencinox
445 745 441 947S 741 741S 746S 946S 441S 446S
139-0101EA 603 EUR
139-0101 139-0102 139-0100 AGEN947S AGEN741 AGEN741S AGEN746S AGEN946S AGEN441S AGEN446S
Trolley, trays
Transportwagen
Trolleys with two handles, shaped trays made up of 18/10 stainless steel with bright polish finishing.
  • Heavy duty structure
  • Easy cleaning
  • Available in knockdown construction or argon welded models