Instrument box, anodised aluminium
Lieferant: Agencinox
9030 9036
AGEN9030EA 52.6 EUR
AGEN9030 AGEN9036
Instrument box, anodised aluminium
Schalen/Schüsseln/Töpfe Instrumentenschalen
Anodised aluminium perforated box.
  • Excellent temperature resistance
  • Good chemical resistance
  • Autoclavable up to 180 °C