TG1 electroporation-competent cells
Lieferant: AGILENT
200123
AGLS200123EA 349 EUR
AGLS200123
TG1 electroporation-competent cells
Kompetente Zellen
Phage display libraries from electroporation-competent cells with high transformation efficiency.
  • Just thaw, combine with DNA, and electroporate to get started quickly
  • Efficiency of ≥1×10¹⁰ transformations/µg of pUC18 DNA
  • Kit size 5×0,1 ml
Electroporation-competent cells capable of producing greater than 1×10¹⁰ transformants/µg of DNA. These electroporation-ready cells need only be thawed, mixed with DNA, and electroporated. TG1 electroporation-competent cells are recommended for preparation of phage display libraries.