Deoxynucleotide mix
Lieferant: AGILENT
The PCR grade deoxynucleotide mix includes 25 mM of each standard dNTP (GATC) for use with any PCR application, including real-time quantitative PCR.
- Ideal for optimal PCR performance
- Produces high-quality reaction products
- Withstands multiple freeze-thaw without compromising efficiency
Lieferumfang: Includes 400 µl of 100 mM dNTP mix (25 mM of each dNTP), adequate for 1000 standard 50 µl primer-extension reactions or 2000 reactions of 25 µl.