QuikChange II
Lieferant: STRATAGENE
These QuikChange II kits provide improved fidelity, while maintaining greater than 80% mutation efficiency for single site mutagenesis.
- Includes PfuUltra High-fidelity DNA polymerase to minimise unwanted errors
- Complete kit provides all reagents necessary to support mutagenesis (at single sites)
Used for large constructs, and for use with electroporation competent cells.
Bitte beachten For research use only. Not for use in diagnostic procedures.