Affinity protein expression and purification systems
Lieferant: STRATAGENE
The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
- pCAL vectors derived from T7 RNA polymerase-based pET vectors
- Novel purification tag combines small size, gentle elution and high-affinity binding
- Easy tag removal resulting in native protein
Lieferumfang: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
Bitte beachten For research use only. Not for use in diagnostic procedures.