Restriction enzymes, Dpn I restriction enzymes
Lieferant: AGILENT
500402
AGLS500402EA 165 EUR
AGLS500402
Restriction enzymes, Dpn I restriction enzymes
Nukleinsäure-Reagenzien Restriktionsenzym
Dpn I is an ideal enzyme for cloning and southern blot analysis.
  • High purity enzyme preparation
Lieferumfang: Includes 10X universal reaction buffer.
Bitte beachten For research use only. Not for use in diagnostic procedures.