Spin concentrators, 50K MWCo
Lieferant: AGILENT
5188-5202
AGLS5188-5202EA 239 EUR
AGLS5188-5202
Spin concentrators, 50K MWCo
Filter Zentrifugaleinheiten
Spin concentrators are devices used to concentrate protein samples via low-speed centrifugation through MWCo filters into a collection tube.
  • Contains a polyethersulfone (PES) membrane that is compatible with biological fluids and most aqueous solutions with pore sizes of 50 kDa
  • Active membrane area of 2,0 or 4,0 cm²
  • Durable and designed to minimise membrane fouling and allow fast concentration, even with particles in the solution
Designed to fit a standard 15 ml conical collection tube or a 50 ml collection tube (not provided with concentrator). Designed to be used in either swing-bucket or fixed angle rotor centrifuge equipment at maximum 5000 or 2000 g.